Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01200389_m1
          See other MBOAT4 GE Assays ›
          Gene Symbol:
          Mboat4
          Gene Name:
          membrane bound O-acyltransferase domain containing 4
          Gene Aliases:
          GOAT, Gm171
          Chromosome Location:
          Chr.8: 34108219 - 34125185 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01200389_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mboat4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001126314.2 NP_001119786.1 2 - 3 410 77
          GenBank mRNA BC147844.1 - 2 - 3 396 9087561 77
          BC147849.1 - 2 - 3 395 9087566 77
          EU518496.1 - 2 - 3 346 77
          EU721729.1 - 2 - 3 410 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000402.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          acetyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-serine octanoylation
          dihydrolipoamide branched chain acyltransferase activity
          sterol O-acyltransferase activity
          N-acetyltransferase activity
          O-acyltransferase activity
          palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
          carnitine O-acyltransferase activity
          acetyltransferase activity
          C-acyltransferase activity
          palmitoyltransferase activity
          N-acyltransferase activity
          acylglycerol O-acyltransferase activity
          serine O-acyltransferase activity
          O-acetyltransferase activity
          O-octanoyltransferase activity
          octanoyltransferase activity
          O-palmitoyltransferase activity
          S-acyltransferase activity
          S-acetyltransferase activity
          S-malonyltransferase activity
          malonyltransferase activity
          C-palmitoyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          succinyltransferase activity
          N-succinyltransferase activity
          O-succinyltransferase activity
          S-succinyltransferase activity
          sinapoyltransferase activity
          O-sinapoyltransferase activity
          peptidyl-lysine N6-myristoyltransferase activity
          peptidyl-lysine N6-palmitoyltransferase activity
          benzoyl acetate-CoA thiolase activity
          3-hydroxybutyryl-CoA thiolase activity
          3-ketopimelyl-CoA thiolase activity
          N-palmitoyltransferase activity
          myristoyltransferase activity
          acyl-CoA N-acyltransferase activity
          protein-cysteine S-myristoyltransferase activity
          protein-cysteine S-acyltransferase activity
          dihydrolipoamide S-acyltransferase activity
          glucosaminyl-phosphotidylinositol O-acyltransferase activity
          ergosterol O-acyltransferase activity
          lanosterol O-acyltransferase activity
          naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
          2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
          2-methylhexanoyl-CoA C-acetyltransferase activity
          butyryl-CoA 2-C-propionyltransferase activity
          2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
          L-2-aminoadipate N-acetyltransferase activity
          UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
          keto acid formate lyase activity
          Ras palmitoyltransferase activity
          azetidine-2-carboxylic acid acetyltransferase activity
          peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
          acetyl-CoA:L-lysine N6-acetyltransferase

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126167 EU721729.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126167 BC147849.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126167 NM_001126314.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126167 BC147844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126167 EU518496.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604717 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274935 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126167 EU721729.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126167 BC147849.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126167 NM_001126314.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126167 BC147844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126167 EU518496.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25772204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06857949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07153287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06915757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07210707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07379314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06794630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07153288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06715836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07006675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07220576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07006829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06794627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06915756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25772250 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10571301 - -
          Affymetrix HT MG-430 PM Array Plate 1439752_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10571300 - -
          Mouse Genome 430 2.0 Array 1439752_at - -
          Mouse Genome 430 2.0 Array 1439752_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800002985.mm.1 NM_001126314.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002985.mm.1 NM_001126314.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002985.mm.1 TC0800000402.mm.1 MTA TC
          Mouse Expression Set 430 1439752_PM_at - -
          Mouse Expression Set 430 1439752_at - -
          Affymetrix HT Mouse Genome 430B Array 1439752_at - -
          Affymetrix HT Mouse Genome 430B Array 1439752_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 344342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline