| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500016404.mm.1 |
TC0500001768.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500016404.mm.1 |
NM_001167944.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500016404.mm.1 |
NM_001167944.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500016404.mm.1 |
NM_016683.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500016404.mm.1 |
NM_016683.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500016405.mm.1 |
TC0500001768.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500016405.mm.1 |
NM_001167944.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500016405.mm.1 |
NM_001167944.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500016405.mm.1 |
NM_016683.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500016405.mm.1 |
NM_016683.2 |
Refseq |