| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500025889.mm.1 |
TC0500002928.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025889.mm.1 |
NR_046195.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025889.mm.1 |
NR_046195.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025890.mm.1 |
TC0500002928.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025890.mm.1 |
NR_046195.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025890.mm.1 |
NR_046195.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025891.mm.1 |
TC0500002928.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025891.mm.1 |
NR_046195.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025891.mm.1 |
NR_046195.1 |
Refseq |