Search
Search

Mlxip| Gene Symbol: Mlxip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133917.3 | NP_598678.2 | 2 - 3 | 647 | 65 | |
| NM_177582.3 | NP_808250.1 | 2 - 3 | 647 | 65 | ||
| GenBank mRNA | AK047475.1 | - | 2 - 3 | 647 | 65 | |
| BC137770.1 | - | 2 - 3 | 574 | 8860789 | 65 | |
Set Membership: |
|