Search
Search

Zdhhc13| Gene Symbol: Zdhhc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028031.3 | NP_082307.1 | 7 - 8 | 855 | 92 | |
| GenBank mRNA | AK010382.1 | - | 7 - 8 | 855 | 92 | |
| AK049640.1 | - | 6 - 7 | 685 | 92 | ||
| AK145855.1 | - | 7 - 8 | 775 | 92 | ||
| AK146196.1 | - | 7 - 8 | 827 | 92 | ||
| BC019493.1 | - | 7 - 8 | 774 | 92 | ||
| BC046599.1 | - | 7 - 8 | 774 | 6311721 | 92 | |
Set Membership: |
|