Search
Search

Wdyhv1| Gene Symbol: Wdyhv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029734.1 | NP_084010.1 | 4 - 5 | 452 | 75 | |
| GenBank mRNA | AK010831.1 | - | 4 - 5 | 473 | 75 | |
| AK150578.1 | - | 4 - 5 | 504 | 75 | ||
| BC051524.1 | - | 4 - 5 | 452 | 1514917 | 75 | |
Set Membership: |
|