| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500013288.mm.1 |
TC0500001466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013288.mm.1 |
NM_001190352.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013288.mm.1 |
NM_001190352.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013284.mm.1 |
TC0500001466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013284.mm.1 |
NM_001190352.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013284.mm.1 |
NM_001190352.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013287.mm.1 |
TC0500001466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013287.mm.1 |
NM_001190352.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013287.mm.1 |
NM_001190352.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013285.mm.1 |
TC0500001466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013285.mm.1 |
NM_001190352.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013285.mm.1 |
NM_001190352.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013286.mm.1 |
TC0500001466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013286.mm.1 |
NM_001190352.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013286.mm.1 |
NM_001190352.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013289.mm.1 |
TC0500001466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013289.mm.1 |
NM_001190352.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013289.mm.1 |
NM_001190352.1 |
Refseq |