Search
Search

Dhx37| Gene Symbol: Dhx37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203319.1 | NP_976064.1 | 6 - 7 | 978 | 60 | |
| GenBank mRNA | AK148257.1 | - | 6 - 7 | 1062 | 60 | |
| AK220399.1 | - | 3 - 4 | 329 | 60 | ||
| BC066077.1 | - | 6 - 7 | 978 | 30354591 | 60 | |
| BC080298.1 | - | 6 - 7 | 996 | 30362983 | 60 | |
Set Membership: |
|