| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10552478 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10552479 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10552476 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10552477 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1453495_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10552475 |
- |
- |
| Mouse Genome 430 2.0 Array |
1453495_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1453495_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
69511_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
69511_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700006258.mm.1 |
TC0700000771.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006258.mm.1 |
NM_027097.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006258.mm.1 |
NM_027097.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006260.mm.1 |
TC0700000771.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006260.mm.1 |
NM_027097.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006260.mm.1 |
NM_027097.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006259.mm.1 |
TC0700000771.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006259.mm.1 |
NM_027097.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006259.mm.1 |
NM_027097.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006262.mm.1 |
TC0700000771.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006262.mm.1 |
NM_027097.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006262.mm.1 |
NM_027097.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006257.mm.1 |
TC0700000771.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006257.mm.1 |
NM_027097.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006257.mm.1 |
NM_027097.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006261.mm.1 |
TC0700000771.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700006261.mm.1 |
NM_027097.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700006261.mm.1 |
NM_027097.1 |
Refseq |
| Mouse Expression Set 430 |
1453495_PM_at |
- |
- |
| Mouse Expression Set 430 |
1453495_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453495_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453495_PM_at |
- |
- |