| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700027116.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027116.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027116.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027120.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027120.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027120.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027122.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027122.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027122.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027133.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027133.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027133.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027134.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027134.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027134.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027123.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027123.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027123.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027124.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027124.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027124.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027131.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027131.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027131.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027132.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027132.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027132.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027139.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027139.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027139.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027140.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027140.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027140.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027144.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027144.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027144.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027148.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027148.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027148.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027110.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027110.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027110.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027137.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027137.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027137.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027121.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027121.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027121.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027149.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027149.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027149.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027150.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027150.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027150.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027115.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027115.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027115.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027119.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027119.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027119.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027138.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027138.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027138.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027141.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027141.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027141.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027117.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027117.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027117.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027118.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027118.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027118.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027135.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027135.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027135.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027142.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027142.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027142.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027151.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027151.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027151.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027125.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027125.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027125.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027126.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027126.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027126.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027127.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027127.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027127.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027128.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027128.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027128.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027130.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027130.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027130.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027145.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027145.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027145.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027111.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027111.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027111.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027136.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027136.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027136.mm.1 |
TC0700003002.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700027143.mm.1 |
NM_029741.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027143.mm.1 |
NM_029741.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700027143.mm.1 |
TC0700003002.mm.1 |
MTA TC |