Search
Search

Sppl3| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029012.2 | NP_083288.2 | 8 - 9 | 848 | 85 | |
| GenBank mRNA | AK014709.1 | - | 8 - 9 | 1186 | 85 | |
| BC023131.1 | - | 4 - 5 | 463 | 85 | ||
| BC086646.1 | - | 8 - 9 | 1213 | 6817386 | 85 | |
| CD806412.1 | - | 4 - 5 | 465 | 85 | ||
Set Membership: |
|