Search
Search

Ankrd13a| Gene Symbol: Ankrd13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026718.2 | NP_080994.2 | 8 - 9 | 977 | 63 | |
| GenBank mRNA | AK039432.1 | - | 8 - 9 | 1172 | 63 | |
| AK156539.1 | - | 8 - 9 | 1170 | 63 | ||
| BC027765.1 | - | 6 - 7 | 649 | 63 | ||
| BC049187.1 | - | 8 - 9 | 1156 | 63 | ||
| BC061692.1 | - | 8 - 9 | 977 | 6308996 | 63 | |
| BC108413.1 | - | 8 - 9 | 1452 | 6487251 | 63 | |
Set Membership: |
|