| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P413147 |
X00472.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P413147 |
X00472.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
348189 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
722232 |
XM_001474864.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
722232 |
XM_001475797.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84672 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P118509 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P413147 |
X00472.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P413147 |
X00472.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P389929 |
X01389.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P389929 |
NM_008693.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P389929 |
BC132657.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P389929 |
BC132659.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25086010 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25086025 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25086029 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25085982 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25086005 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25085988 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25086043 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25086020 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25086030 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P118509 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P413147 |
X00472.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P413147 |
X00472.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P111106 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P118509 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06096268 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06096262 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05932058 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25085982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086084 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086116 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25085968 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086113 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086033 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086060 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086094 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25085948 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086043 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086005 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086034 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086042 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086073 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086038 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086036 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25085979 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086115 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086063 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086088 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086090 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086029 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25086062 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06555500 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06010192 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25085982 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086025 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086020 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086036 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086010 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25085979 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086029 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086042 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05920917 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086038 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25085988 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06555512 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086030 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086043 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06096268 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25086005 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06342465 |
- |
- |