Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01203945_m1
          See other LIPN GE Assays ›
          Gene Symbol:
          Lipn
          Gene Name:
          lipase, family member N
          Gene Aliases:
          2210418G03Rik, Lipl4
          Chromosome Location:
          Chr.19: 34067058 - 34085946 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01203945_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lipn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027340.2 NP_081616.1 7 - 8 1058 67
          GenBank mRNA AK008974.1 - 6 - 7 688 67
          AK154333.1 - 7 - 8 1059 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000491.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          hydrolase lipase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          biological_process
          lipid catabolic process
          molecular_function
          phospholipase activity
          prenylcysteine methylesterase activity
          lipase activity
          hydrolase activity
          hydrolase activity, acting on ester bonds
          1-oxa-2-oxocycloheptane lactonase activity
          sulfolactone hydrolase activity
          3,4-dihydrocoumarin hydrolase activity
          butyrolactone hydrolase activity
          endosulfan lactone lactonase activity
          L-ascorbate 6-phosphate lactonase activity
          Ser-tRNA(Thr) hydrolase activity
          Ala-tRNA(Pro) hydrolase activity
          Cys-tRNA(Pro) hydrolase activity
          Ser(Gly)-tRNA(Ala) hydrolase activity
          all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
          retinyl-palmitate esterase activity
          mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
          mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
          methyl indole-3-acetate esterase activity
          methyl salicylate esterase activity
          methyl jasmonate esterase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057430 AK154333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057430 NM_027340.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14025 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548756 AK035423.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548756 BC049605.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548756 NM_011321.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 705848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 154118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91914 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110869 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157394 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168335 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057430 AK154333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057430 NM_027340.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110869 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157394 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168335 - -
          Whole Mouse Genome G4122A A_51_P465386 AK154333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P465386 NM_027340.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P465386 AK008974.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157394 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110869 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32078190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14424663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14363609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14269887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14269884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14465345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14423567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14465349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14498613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14269891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14379708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14424664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32078095 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10462574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462570 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462568 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462571 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462569 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462567 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462572 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462566 - -
          Affymetrix HT MG-430 PM Array Plate 1431492_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10462565 - -
          Mouse Genome 430 2.0 Array 1431492_at - -
          Mouse Genome 430 2.0 Array 1431492_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70166_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70166_at - -
          Affymetrix Mouse Clariom S Array PSR1900004389.mm.1 NM_027340.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004389.mm.1 NM_027340.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004389.mm.1 TC1900000491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004390.mm.1 NM_027340.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004390.mm.1 NM_027340.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004390.mm.1 TC1900000491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004387.mm.1 NM_027340.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004387.mm.1 NM_027340.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004387.mm.1 TC1900000491.mm.1 MTA TC
          Mouse Expression Set 430 1431492_PM_at - -
          Mouse Expression Set 430 1431492_at - -
          Affymetrix HT Mouse Genome 430B Array 1431492_at - -
          Affymetrix HT Mouse Genome 430B Array 1431492_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 298882 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2944988 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2944988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline