Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01204485_s1
          See other VGF GE Assays ›
          Gene Symbol:
          Vgf
          Gene Name:
          VGF nerve growth factor inducible
          Gene Aliases:
          Gm1052
          Chromosome Location:
          Chr.5: 137025302 - 137033351 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01204485_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vgf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039385.1 NP_001034474.1 2 2066 125
          GenBank mRNA BC085134.1 - 5 2347 6734388 125
          BC138553.1 - 1 1912 8861573 125
          BC138554.1 - 1 1912 8861574 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          response to dietary excess
          generation of precursor metabolites and energy
          response to cold
          sexual reproduction
          insulin secretion
          response to insulin
          glucose homeostasis
          penile erection
          regulation of neuronal synaptic plasticity
          neuropeptide hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124216 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124211 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124233 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124213 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124223 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124231 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124227 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085060 NM_001039385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085060 BC085134.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885629 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71568 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085060 NM_001039385.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085060 BC085134.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124228 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05759645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124223 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124231 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124233 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124216 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124213 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124211 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04553062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04680170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04281721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04281716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04680160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04229072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04207140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04680169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04207144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04381994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04680162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04281717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04381996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04566405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04680167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04551799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04207149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124198 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526555 - -
          Affymetrix HT MG-430 PM Array Plate 1436094_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526553 - -
          Mouse Genome 430 2.0 Array 1436094_PM_at - -
          Mouse Genome 430 2.0 Array 1436094_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381677_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381677_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500014648.mm.1 TC0500001618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014645.mm.1 TC0500001618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014647.mm.1 TC0500001618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014650.mm.1 TC0500001618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014653.mm.1 TC0500001618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014653.mm.1 NM_001039385.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014653.mm.1 NM_001039385.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014654.mm.1 TC0500001618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014654.mm.1 NM_001039385.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014654.mm.1 NM_001039385.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014651.mm.1 TC0500001618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014651.mm.1 NM_001039385.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014651.mm.1 NM_001039385.1 Refseq
          Mouse Expression Set 430 1436094_at - -
          Mouse Expression Set 430 1436094_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436094_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436094_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 448669 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232310 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243040 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2925923 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2925923 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0