Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Plod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011962.3 | NP_036092.1 | 7 - 8 | 1452 | 118 | |
GenBank mRNA | AF046783.1 | - | 7 - 8 | 860 | 118 | |
AK013195.1 | - | 7 - 8 | 998 | 118 | ||
AK033360.1 | - | 7 - 8 | 997 | 118 | ||
AK053320.1 | - | 7 - 8 | 1094 | 118 | ||
AK088948.1 | - | 7 - 8 | 869 | 118 | ||
AK157741.1 | - | 7 - 8 | 961 | 118 | ||
BC043047.1 | - | 7 - 8 | 1451 | 5703967 | 118 | |
BC054734.1 | - | 7 - 8 | 1452 | 5720686 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |