| Array Name |
Probe IDs |
Targets |
Target Type |
| Mouse Genome 430 2.0 Array |
1437555_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1437555_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0500025686.mm.1 |
TC0500002910.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025686.mm.1 |
NM_001005477.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025686.mm.1 |
NM_001005477.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025685.mm.1 |
TC0500002910.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025685.mm.1 |
NM_001005477.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025685.mm.1 |
NM_001005477.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025684.mm.1 |
TC0500002910.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025684.mm.1 |
NM_001005477.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025684.mm.1 |
NM_001005477.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025687.mm.1 |
TC0500002910.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025687.mm.1 |
NM_001005477.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025687.mm.1 |
NM_001005477.1 |
Refseq |
| Mouse Expression Set 430 |
1437555_at |
- |
- |
| Mouse Expression Set 430 |
1437555_PM_at |
- |
- |