Search
Search

Ddx51| Gene Symbol: Ddx51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027156.3 | NP_081432.2 | 4 - 5 | 769 | 60 | |
| GenBank mRNA | AK152599.1 | - | 4 - 5 | 737 | 60 | |
| BC060646.1 | - | 4 - 5 | 746 | 6853826 | 60 | |
Set Membership: |
|