Search
Search

Oplah| Gene Symbol: Oplah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153122.2 | NP_694762.1 | 15 - 16 | 2171 | 57 | |
| GenBank mRNA | AK158391.1 | - | 15 - 16 | 2213 | 57 | |
| BC034522.1 | - | 15 - 16 | 2171 | 4236444 | 57 | |
Set Membership: |
|