Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01205601_g1
          See other PPP1R15A GE Assays ›
          Gene Symbol:
          Ppp1r15a
          Gene Name:
          protein phosphatase 1, regulatory (inhibitor) subunit 15A
          Gene Aliases:
          9630030H21, Gadd34, Myd116
          Chromosome Location:
          Chr.7: 45522917 - 45526268 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01205601_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r15a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008654.2 NP_032680.1 2 - 3 1925 75
          GenBank mRNA AK136336.1 - 2 - 3 1881 75
          AK154697.1 - 1 - 2 2407 75
          AK167317.1 - 3 - 4 1650 75
          BC138485.1 - 2 - 3 1790 8861505 75
          BC138486.1 - 2 - 3 1790 8861506 75
          X51829.1 - 2 - 3 1870 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003010.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r15a

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          apoptotic process
          positive regulation of gene expression
          endoplasmic reticulum unfolded protein response
          negative regulation of phosphoprotein phosphatase activity
          positive regulation of phosphoprotein phosphatase activity
          positive regulation of peptidyl-serine phosphorylation
          response to endoplasmic reticulum stress
          negative regulation of protein dephosphorylation
          regulation of translational initiation by eIF2 alpha dephosphorylation
          positive regulation of transcription from RNA polymerase I promoter
          regulation of catalytic activity
          regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          peptidyl-serine dephosphorylation
          protein localization to endoplasmic reticulum
          positive regulation of peptidyl-serine dephosphorylation
          negative regulation of response to endoplasmic reticulum stress
          negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          protein phosphatase 1 binding
          protein phosphatase regulator activity
          protein kinase binding
          protein phosphatase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093636 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136880 BC138486.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136880 X51829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136880 AK167317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136880 AW493042.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136880 BC138485.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101566 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175412 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136880 BC138486.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136880 X51829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136880 AK167317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136880 AW493042.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136880 BC138485.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25093608 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175412 - -
          Whole Mouse Genome G4122A A_51_P302503 BC138486.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302503 X51829.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302503 AK167317.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302503 BC138485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302503 AK136336.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132099 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125358 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113899 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175412 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06154697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06154702 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06154709 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06154689 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06154700 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06154692 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093636 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25093639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06141601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06141602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06154689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06154691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25093626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563341 - -
          Affymetrix HT MG-430 PM Array Plate 1448325_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563338 - -
          Mouse Genome 430 2.0 Array 1448325_PM_at - -
          Mouse Genome 430 2.0 Array 1448325_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17872_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17872_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700027303.mm.1 NM_008654.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027303.mm.1 NM_008654.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027303.mm.1 TC0700003010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027304.mm.1 NM_008654.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027304.mm.1 NM_008654.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027304.mm.1 TC0700003010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027305.mm.1 NM_008654.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027305.mm.1 NM_008654.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027305.mm.1 TC0700003010.mm.1 MTA TC
          Mouse Expression Set 430 1448325_PM_at - -
          Mouse Expression Set 430 1448325_at - -
          Murine Genome U74v2 Set 160463_PM_at - -
          Murine Genome U74v2 Set 160463_at - -
          Mouse Genome 430A 2.0 Array 1448325_at - -
          Mouse Genome 430A 2.0 Array 1448325_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448325_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448325_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6350601 - -
          Illumina mouse-6 v1.0 expression beadchip 6350601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350601 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2722938 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2722938 - -
          mouse-6 v1.1 (Illumina) scl31371.4.1_45-S - -
          Illumina Sentrix mouse-6.v1 bead array 6350601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31371.4.1_45-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350601 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2722938 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2722938 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline