Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01207064_m1
          See other MUC3 GE Assays ›
          Gene Symbol:
          Muc3
          Gene Name:
          mucin 3, intestinal
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 137134924 - 137166048 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01207064_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Muc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF027131.1 - 10 - 11 1290 110
          AK132919.1 - 4 - 5 433 110
          AK136468.1 - 4 - 5 489 110
          BC015298.1 - 9 - 10 1136 110
          BC026665.1 - 9 - 10 972 110
          BC046639.1 - 8 - 9 877 110
          BC058768.1 - 8 - 9 819 110

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 BC058768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 BC046639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 AK132919.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 AK136468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 BC015298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 BC058768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 BC046639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 AK132919.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 AK136468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450406 BC015298.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132721 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124757 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 BC058768.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 BC046639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 AK132919.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 AK136468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 BC015298.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 BC058768.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 BC046639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 AK132919.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 AK136468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450406 BC015298.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05759919 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124757 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132721 - -
          Whole Mouse Genome G4122A A_52_P536407 BC046639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536407 BC015298.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536407 BC026665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536407 AF027131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 BC058768.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 BC046639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 AK132919.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 AK136468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 BC015298.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 BC058768.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 BC046639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 AK132919.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 AK136468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450406 BC015298.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139209 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132721 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04794141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04356239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04350667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04150484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04903847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04371558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04469592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04971289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04116547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04968285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04371560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04794140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04903846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04971287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04662129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04116548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04350669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04315570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04539226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04697239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124799 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534691 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534690 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534685 - -
          Affymetrix HT MG-430 PM Array Plate 1427374_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534684 - -
          Mouse Genome 430 2.0 Array 1427374_at - -
          Mouse Genome 430 2.0 Array 1427374_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 666339_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 666339_PM_at - -
          Mouse Expression Set 430 1427374_PM_at - -
          Mouse Expression Set 430 1427374_at - -
          Murine Genome U74v2 Set 101390_at - -
          Murine Genome U74v2 Set 101390_PM_at - -
          Mouse Genome 430A 2.0 Array 1427374_at - -
          Mouse Genome 430A 2.0 Array 1427374_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427374_at - -
          Affymetrix HT Mouse Genome 430A Array 1427374_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 775046 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101690750 - -
          Illumina mouse-6 v1.0 expression beadchip 105670541 - -
          Illumina mouse-6 v1.0 expression beadchip 101690750 - -
          mouse-6 v1.1 (Illumina) GI_38082355-S - -
          mouse-6 v1.1 (Illumina) scl25888.8.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 101690750 - -
          Illumina Sentrix mouse-6.v1 bead array 105670541 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105670541 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101690750 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235506 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2718570 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0