| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500027705.mm.1 |
TC0500003111.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027705.mm.1 |
NM_001101546.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027705.mm.1 |
NM_001101546.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027706.mm.1 |
TC0500003111.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027706.mm.1 |
NM_001101546.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027706.mm.1 |
NM_001101546.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027707.mm.1 |
TC0500003111.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027707.mm.1 |
NM_001101546.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027707.mm.1 |
NM_001101546.1 |
Refseq |