Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01207256_s1
          See other OLFR1426 GE Assays ›
          Gene Symbol:
          Olfr1426
          Gene Name:
          olfactory receptor 1426
          Gene Aliases:
          MOR239-1
          Chromosome Location:
          Chr.19: 12087610 - 12091847 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01207256_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1426
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146809.2 NP_667020.1 3 1118 97
          GenBank mRNA BC058600.1 - 3 1118 6390368 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1426

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087547 NM_146809.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087547 BC058600.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81824 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119152 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115824 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087547 NM_146809.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087547 BC058600.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P457759 NM_146809.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P457759 BC058600.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31959465 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115824 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09107935 - -
          Whole Mouse Genome G4122A A_51_P457759 NM_146809.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P457759 BC058600.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121841 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128527 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116762 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115824 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14474999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14474993 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14474991 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14474997 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14475002 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14474990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31959449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14387839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14387841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14387826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14158472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14475005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14280287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14196707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14321125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31959465 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466284 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466280 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466282 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1770239 - -
          Illumina mouse-6 v1.0 expression beadchip 1770239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2624919 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2624919 - -
          mouse-6 v1.1 (Illumina) scl52713.3_469-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770239 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52713.3_469-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2906440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2624919 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2624919 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2906440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0