Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01207311_s1
          See other OLFR1441 GE Assays ›
          Gene Symbol:
          Olfr1441
          Gene Name:
          olfactory receptor 1441
          Gene Aliases:
          MOR215-2
          Chromosome Location:
          Chr.19: 12422311 - 12423267 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01207311_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1441
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146683.1 NP_666894.1 1 906 97
          GenBank mRNA BC145847.1 - 1 966 40131150 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1441

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147315 NM_146683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147315 BC145847.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267288 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157241 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147315 NM_146683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147315 BC145847.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P318354 NM_146683.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P318354 BC145847.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31960616 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157241 - -
          Whole Mouse Genome G4122A A_52_P318354 NM_146683.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P318354 BC145847.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132001 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157241 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14263233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14263225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31960627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14479646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14479640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14420069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14406029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14263232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14406028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14409206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31960653 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461716 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461715 - -
          Affymetrix Mouse Clariom S Array PSR1900002895.mm.1 TC1900000243.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 770537 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5080369 - -
          Illumina mouse-6 v1.0 expression beadchip 5080369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080369 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222683 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222683 - -
          mouse-6 v1.1 (Illumina) scl53345.1.1_224-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080369 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53345.1.1_224-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080369 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222683 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222683 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53345.1.1_224-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0