Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01207595_s1
          See other OLFR1419 GE Assays ›
          Gene Symbol:
          Olfr1419
          Gene Name:
          olfactory receptor 1419
          Gene Aliases:
          MOR266-10
          Chromosome Location:
          Chr.19: 11870183 - 11871214 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01207595_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1419
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011775.1 NP_001011775.1 1 981 97
          GenBank mRNA BC147200.1 - 1 1072 9056244 97
          BC147201.1 - 1 1072 9056264 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1419

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31958228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31958227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31958229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31958296 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232438 NM_001011775.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232438 BC147200.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232438 BC147201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232438 NM_001011775.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232438 BC147200.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P232438 BC147201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232438 NM_001011775.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232438 BC147200.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232438 BC147201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232438 NM_001011775.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232438 BC147200.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P232438 BC147201.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232438 NM_001011775.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232438 BC147200.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232438 BC147201.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232438 NM_001011775.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232438 BC147200.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P232438 BC147201.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958265 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03551164 - -
          Whole Mouse Genome G4122A A_52_P232438 NM_001011775.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232438 BC147200.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232438 BC147201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232438 NM_001011775.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232438 BC147200.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P232438 BC147201.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14232202 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14232206 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14430359 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14430349 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14430353 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14232205 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14430355 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31958227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31958228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31958296 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31958229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14232202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14412237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14458447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14175310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14267871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14462318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14433851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958236 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466273 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466272 - -
          Affymetrix Mouse Clariom S Array PSR1900010753.mm.1 TC1900001147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010754.mm.1 NM_001011775.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010754.mm.1 NM_001011775.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010754.mm.1 TC1900001147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010755.mm.1 TC1900001147.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 410640 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161272 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161272 - -
          mouse-6 v1.1 (Illumina) GI_58801325-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161272 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161272 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0