Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01207636_m1
          See other SBDS GE Assays ›
          Gene Symbol:
          Sbds
          Gene Name:
          Shwachman-Bodian-Diamond syndrome homolog (human)
          Gene Aliases:
          4733401P19Rik, AI836084, CGI-97, SDS
          Chromosome Location:
          Chr.5: 130245731 - 130255462 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01207636_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sbds
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023248.1 NP_075737.1 4 - 5 754 108
          GenBank mRNA AK005079.1 - 4 - 5 731 108
          AK014648.1 - 4 - 5 759 108
          AK019228.1 - 4 - 5 741 108
          AK168847.1 - 4 - 5 741 108
          BC003849.1 - 4 - 5 754 3599733 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003318.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sbds

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          inner cell mass cell proliferation
          rRNA processing
          mitotic spindle organization
          cell proliferation
          bone mineralization
          leukocyte chemotaxis
          ribosome biogenesis
          mature ribosome assembly
          bone marrow development
          RNA binding
          microtubule binding
          rRNA binding
          ribosome binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24092945 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24092948 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24092947 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 AK014648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 AK168847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 BC003849.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 NM_023248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 AK005079.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 AK014648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 AK168847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 BC003849.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 NM_023248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466875 AK005079.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533728 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369201 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174573 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100285 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 AK014648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 AK168847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 BC003849.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 NM_023248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 AK005079.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 AK014648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 AK168847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 BC003849.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 NM_023248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466875 AK005079.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24092943 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100285 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01220900 - -
          Whole Mouse Genome G4122A A_51_P466875 AK014648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466875 AK168847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466875 BC003849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466875 NM_023248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466875 AK005079.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466875 AK014648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466875 AK168847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466875 BC003849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466875 NM_023248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466875 AK005079.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178738 AK014648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178738 AK019228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178738 AK168847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178738 BC003849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178738 NM_023248.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138507 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100285 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04197674 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24092948 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24092945 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24092947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24092893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04906617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04236442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04906618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04124063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04610139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04511181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04124064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04791864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24092910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04236436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04860617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04511188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04124062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04236437 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534143 - -
          Affymetrix HT MG-430 PM Array Plate 1426480_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534142 - -
          Mouse Genome 430 2.0 Array 1426480_at - -
          Mouse Genome 430 2.0 Array 1426480_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66711_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66711_at - -
          Affymetrix Mouse Clariom S Array PSR0500030040.mm.1 TC0500003318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030040.mm.1 NM_023248.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030039.mm.1 TC0500003318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030039.mm.1 NM_023248.2 Refseq
          Mouse Expression Set 430 1426480_at - -
          Mouse Expression Set 430 1426480_PM_at - -
          Murine Genome U74v2 Set 95752_at - -
          Murine Genome U74v2 Set 95752_PM_at - -
          Mouse Genome 430A 2.0 Array 1426480_at - -
          Mouse Genome 430A 2.0 Array 1426480_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426480_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426480_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 469345 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4120441 - -
          Illumina mouse-6 v1.0 expression beadchip 4120441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004120441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245650 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245650 - -
          mouse-6 v1.1 (Illumina) scl25991.5.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 4120441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25991.5.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245650 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245650 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl25991.5.1_3-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline