Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01208549_m1
          See other SCD4 GE Assays ›
          Gene Symbol:
          Scd4
          Gene Name:
          stearoyl-coenzyme A desaturase 4
          Gene Aliases:
          ACOD4
          Chromosome Location:
          Chr.19: 44333326 - 44346743 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01208549_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183216.3 NP_899039.2 5 - 6 1030 63
          GenBank mRNA AY430080.1 - 5 - 6 1030 63
          BC141234.1 - 5 - 6 986 9055846 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000647.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scd4

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid biosynthetic process
          oxidation-reduction process
          monounsaturated fatty acid biosynthetic process
          stearoyl-CoA 9-desaturase activity
          palmitoyl-CoA 9-desaturase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011872 AY430080.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011872 NM_183216.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 91886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 31202 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136857 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180152 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011872 AY430080.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011872 NM_183216.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32131697 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180152 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09180909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03603078 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136857 - -
          Whole Mouse Genome G4122A A_52_P603237 BC038322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P374877 AY430080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P374877 NM_183216.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136857 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180152 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14400871 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14400910 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14400904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14400876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32131750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14286208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14400893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14502755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14400900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14400865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14400884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14443338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14286209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14400913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14180566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14449178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32131684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14400897 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10463365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463367 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463368 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463370 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463369 - -
          Affymetrix Mouse Gene 1.1 ST Array 10463361 - -
          Affymetrix Mouse Clariom S Array PSR1900005736.mm.1 NM_183216.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005736.mm.1 NM_183216.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005736.mm.1 TC1900000647.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005734.mm.1 NM_183216.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005734.mm.1 NM_183216.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005734.mm.1 TC1900000647.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005735.mm.1 NM_183216.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005735.mm.1 NM_183216.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005735.mm.1 TC1900000647.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 738720 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220332 - -
          Illumina mouse-6 v1.0 expression beadchip 7040278 - -
          Illumina mouse-6 v1.0 expression beadchip 5220332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2653871 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259674 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2653871 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259674 - -
          mouse-6 v1.1 (Illumina) scl0329065.6_75-S - -
          mouse-6 v1.1 (Illumina) scl53097.1.2_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220332 - -
          Illumina Sentrix mouse-6.v1 bead array 7040278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53097.1.2_25-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0329065.6_75-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259674 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2653871 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259674 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2653871 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline