Search
Search
| Gene Symbol: Ldb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039071.2 | NP_001034160.1 | 2 - 3 | 198 | 59 | |
| NM_001039072.2 | NP_001034161.1 | 2 - 3 | 198 | 59 | ||
| NM_001039073.2 | NP_001034162.1 | 2 - 3 | 198 | 59 | ||
| NM_001039074.2 | NP_001034163.1 | 1 - 2 | 229 | 59 | ||
| NM_001039075.2 | NP_001034164.1 | 2 - 3 | 198 | 59 | ||
| NM_001039076.2 | NP_001034165.1 | 2 - 3 | 198 | 59 | ||
| NM_011918.4 | NP_036048.3 | 2 - 3 | 198 | 59 | ||
| GenBank mRNA | AF114378.2 | - | 2 - 3 | 164 | 59 | |
| AF114379.2 | - | 2 - 3 | 162 | 59 | ||
| AF228057.1 | - | 1 - 2 | 91 | 59 | ||
| AF228058.1 | - | 1 - 2 | 91 | 59 | ||
| AJ005621.1 | - | 2 - 3 | 191 | 59 | ||
| AK004020.1 | - | 2 - 3 | 206 | 59 | ||
| AK084461.1 | - | 2 - 3 | 178 | 59 | ||
| AK137181.1 | - | 2 - 3 | 181 | 59 | ||
| AK142292.1 | - | 2 - 3 | 150 | 59 | ||
| AK172980.1 | - | 1 - 2 | 229 | 59 | ||
| AY206011.1 | - | 1 - 2 | 115 | 59 | ||
| AY206012.1 | - | 2 - 3 | 164 | 59 | ||
| AY206013.1 | - | 2 - 3 | 162 | 59 | ||
| AY206015.1 | - | 1 - 2 | 114 | 59 | ||
| BC099596.1 | - | 2 - 3 | 165 | 59 | ||
| BC138793.1 | - | 1 - 2 | 132 | 8861815 | 59 | |
| BC145420.1 | - | 1 - 2 | 132 | 9053978 | 59 | |
| BY704269.1 | - | 2 - 3 | 205 | 59 | ||
Set Membership: |
|