Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01208811_m1
          See other GM5938 GE Assays ›
          Gene Symbol:
          Gm5938
          Gene Name:
          predicted gene 5938
          Gene Aliases:
          EG546335, OBP6
          Chromosome Location:
          Chr.X: 78125467 - 78130403 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01208811_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5938
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085534.2 NP_001079003.1 4 - 5 426 96
          GenBank mRNA CK615904.1 - 4 - 5 397 96
          CK616445.1 - 4 - 5 426 96
          CK616524.1 - 3 - 4 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000782.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987953 NM_001085534.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225315 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987953 NM_001085534.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32480434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32480411 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00005289.mm.1 NM_001085534.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005289.mm.1 TC0X00000782.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005290.mm.1 NM_001085534.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005290.mm.1 TC0X00000782.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005293.mm.1 NM_001085534.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005293.mm.1 TC0X00000782.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005296.mm.1 NM_001085534.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005296.mm.1 TC0X00000782.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005294.mm.1 NM_001085534.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005294.mm.1 TC0X00000782.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005297.mm.1 NM_001085534.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005297.mm.1 TC0X00000782.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005292.mm.1 NM_001085534.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005292.mm.1 TC0X00000782.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline