Search
Search

Cars2| Gene Symbol: Cars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024248.1 | NP_077210.1 | 9 - 10 | 1039 | 100 | |
| GenBank mRNA | AK009937.1 | - | 9 - 10 | 1001 | 100 | |
| AK038968.1 | - | 9 - 10 | 1049 | 100 | ||
Set Membership: |
|