Search Thermo Fisher Scientific
Gene Symbol: Klhl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290477.1 | NP_001277406.1 | 4 - 5 | 1016 | 87 | |
NM_001290478.1 | NP_001277407.1 | 4 - 5 | 1016 | 87 | ||
NM_172781.2 | NP_766369.2 | 4 - 5 | 1016 | 87 | ||
GenBank mRNA | AK044930.1 | - | 4 - 5 | 747 | 87 | |
AK081448.1 | - | 4 - 5 | 1017 | 87 | ||
AK081937.1 | - | 4 - 5 | 829 | 87 | ||
AK085099.1 | - | 4 - 5 | 1017 | 87 | ||
AK161786.1 | - | 4 - 5 | 910 | 87 | ||
BC057137.1 | - | 4 - 5 | 747 | 87 | ||
BC099954.1 | - | 4 - 5 | 843 | 30081872 | 87 | |
BC141251.1 | - | 4 - 5 | 759 | 9055863 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |