Search
Search

4930474N05Rik| Gene Symbol: 4930474N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175008.3 | NP_778173.1 | 1 - 2 | 206 | 72 | |
| GenBank mRNA | AK076810.1 | - | 1 - 2 | 207 | 72 | |
| AK132960.1 | - | 215 | 72 | |||
| BC096580.1 | - | 1 - 2 | 263 | 3470874 | 72 | |
| BC132442.1 | - | 1 - 2 | 202 | 40130719 | 72 | |
| BC132446.1 | - | 1 - 2 | 202 | 40130723 | 72 | |
Set Membership: |
|