Search
Search

Tacc1| Gene Symbol: Tacc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177089.5 | NP_796063.3 | 10 - 11 | 2296 | 69 | |
| NM_199323.3 | NP_955355.1 | 8 - 9 | 1072 | 69 | ||
| GenBank mRNA | AK158441.1 | - | 10 - 11 | 2303 | 69 | |
| AY177412.1 | - | 8 - 9 | 980 | 69 | ||
| AY177413.1 | - | 10 - 11 | 2198 | 69 | ||
| BC125389.1 | - | 8 - 9 | 882 | 69 | ||
| BC145709.1 | - | 8 - 9 | 882 | 69 | ||
Set Membership: |
|