Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01210323_m1
          See other ABLIM2 GE Assays ›
          Gene Symbol:
          Ablim2
          Gene Name:
          actin-binding LIM protein 2
          Gene Aliases:
          AI606905, C230091L11
          Chromosome Location:
          Chr.5: 35757855 - 35884979 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01210323_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ablim2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177696.1 NP_001171167.1 5 - 6 758 72
          NM_001177697.1 NP_001171168.1 5 - 6 758 72
          NM_001177698.1 NP_001171169.1 5 - 6 758 72
          NM_001177699.1 NP_001171170.1 5 - 6 758 72
          NM_001177700.1 NP_001171171.1 5 - 6 758 72
          NM_177678.7 NP_808346.3 5 - 6 758 72
          GenBank mRNA AJ748602.1 - 5 - 6 714 72
          AJ748603.1 - 5 - 6 714 72
          AK046243.1 - 5 - 6 709 72
          AK046879.1 - 5 - 6 707 72
          AK082707.1 - 5 - 6 708 72
          AK154494.1 - 5 - 6 760 72
          AY274116.1 - 5 - 6 685 72
          BC141125.1 - 5 - 6 618 9055737 72
          DQ413175.1 - 4 - 5 587 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000400.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ablim2

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          cytoskeleton organization
          actin cytoskeleton organization
          positive regulation of transcription from RNA polymerase II promoter
          molecular_function
          actin binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 TC0500000400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177696.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177696.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177697.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177697.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177698.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177698.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177699.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177699.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177700.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_001177700.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_177678.7.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500003989.mm.1 NM_177678.7 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0