| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600015812.mm.1 |
NM_001045515.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600015812.mm.1 |
NM_001045515.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600015812.mm.1 |
NM_001164483.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600015812.mm.1 |
NM_001164483.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600015812.mm.1 |
TC1600002060.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600015814.mm.1 |
NM_001045515.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600015814.mm.1 |
NM_001045515.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600015814.mm.1 |
NM_001164483.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600015814.mm.1 |
NM_001164483.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600015814.mm.1 |
TC1600002060.mm.1 |
MTA TC |