Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01210799_m1
          See other GAREML GE Assays ›
          Gene Symbol:
          Gareml
          Gene Name:
          GRB2 associated, regulator of MAPK1-like
          Gene Aliases:
          Fam59b, Gm444
          Chromosome Location:
          Chr.5: 30105196 - 30118380 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01210799_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gareml
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167879.1 NP_001161351.1 1 - 2 205 96
          GenBank mRNA CX204487.1 - 1 - 2 188 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000285.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957053 CJ217704.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957053 NM_001167879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957053 AK029017.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957053 NM_178878.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038854 BC113783.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038854 BC113782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038852 BC113783.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038852 BC113782.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119385 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157679 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957053 CJ217704.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957053 NM_001167879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957053 AK029017.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957053 NM_178878.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157679 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119385 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119385 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04305818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23570742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23570739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04426188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23570735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04943743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04426187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23570740 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10520460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520464 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520462 - -
          Affymetrix HT MG-430 PM Array Plate 1439570_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10520459 - -
          Mouse Genome 430 2.0 Array 1439570_at - -
          Mouse Genome 430 2.0 Array 1439570_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242915_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242915_at - -
          Affymetrix Mouse Clariom S Array PSR0500002181.mm.1 TC0500000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002181.mm.1 NM_001167879.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002181.mm.1 NM_001167879.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002183.mm.1 TC0500000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002183.mm.1 NM_001167879.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002183.mm.1 NM_001167879.1 Refseq
          Mouse Expression Set 430 1439570_at - -
          Mouse Expression Set 430 1439570_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439570_at - -
          Affymetrix HT Mouse Genome 430B Array 1439570_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250997 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline