Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01210983_m1
          See other HTRA4 GE Assays ›
          Gene Symbol:
          Htra4
          Gene Name:
          HtrA serine peptidase 4
          Gene Aliases:
          B430206E18Rik
          Chromosome Location:
          Chr.8: 25024928 - 25038962 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01210983_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htra4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081187.3 NP_001074656.1 2 - 3 671 58
          GenBank mRNA AK046614.1 - 2 - 3 590 58
          BC132380.1 - 2 - 3 631 40130657 58
          BC145842.1 - 2 - 3 631 40131145 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001902.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          proteolysis
          endopeptidase activity
          serine-type endopeptidase activity
          insulin-like growth factor binding
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P207314 BC132380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P207314 BC145842.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P207314 NM_001081187.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P207314 AK046614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P207314 BC132380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P207314 BC145842.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P207314 NM_001081187.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P207314 AK046614.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464403 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358097 AK036575.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358097 AK042661.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358097 AK129217.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358097 AK140214.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358097 NM_133227.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34029 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P207314 BC132380.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P207314 BC145842.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P207314 NM_001081187.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P207314 AK046614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P207314 BC132380.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P207314 BC145842.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P207314 NM_001081187.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P207314 AK046614.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01694558 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01694550 - -
          Whole Mouse Genome G4122A A_52_P207314 BC132380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P207314 BC145842.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P207314 NM_001081187.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P207314 AK046614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P207314 BC132380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P207314 BC145842.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P207314 NM_001081187.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P207314 AK046614.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25725004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25725001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25724831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07033180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06681775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06919045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07251268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07340844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25725001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06987597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07257349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07257339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25725004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07288369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06919047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25724946 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577791 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577788 - -
          Affymetrix HT MG-430 PM Array Plate 1437273_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10577782 - -
          Mouse Genome 430 2.0 Array 1437273_at - -
          Mouse Genome 430 2.0 Array 1437273_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 330723_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 330723_at - -
          Affymetrix Mouse Clariom S Array PSR0800015172.mm.1 NM_001081187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015172.mm.1 NM_001081187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015172.mm.1 TC0800001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800015174.mm.1 NM_001081187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015174.mm.1 NM_001081187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015174.mm.1 TC0800001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800015175.mm.1 NM_001081187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015175.mm.1 NM_001081187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015175.mm.1 TC0800001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800015176.mm.1 NM_001081187.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015176.mm.1 NM_001081187.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015176.mm.1 TC0800001902.mm.1 MTA TC
          Mouse Expression Set 430 1437273_PM_at - -
          Mouse Expression Set 430 1437273_at - -
          Murine Genome U74v2 Set 129661_at - -
          Murine Genome U74v2 Set 129661_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437273_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437273_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 766991 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2669289 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2669289 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0