Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01211129_m1
          See other NOM1 GE Assays ›
          Gene Symbol:
          Nom1
          Gene Name:
          nucleolar protein with MIF4G domain 1
          Gene Aliases:
          D5Kng1, Gm1040
          Chromosome Location:
          Chr.5: 29434659 - 29452169 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01211129_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nom1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033457.2 NP_001028629.2 10 - 11 2407 66
          GenBank mRNA AK148406.1 - 10 - 11 2411 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000272.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Process(es)

          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567505 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567511 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107255 NM_001033457.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P107255 AK148406.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80303 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84722 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113479 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107255 NM_001033457.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P107255 AK148406.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23567515 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05517907 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113479 - -
          Whole Mouse Genome G4122A A_52_P336713 NM_001033457.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P336713 AK148406.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P87001 NM_001033457.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P87001 AK148406.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05517907 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113479 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23567505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23567660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04928821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04167057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04502934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04449822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04169306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04502938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04449828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04502951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04528730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04586223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04989560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04449837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04528728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04989559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23567545 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10520406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520407 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520416 - -
          Affymetrix Mouse Gene 1.1 ST Array 10520405 - -
          Affymetrix Mouse Clariom S Array PSR0500002073.mm.1 TC0500000272.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002073.mm.1 NM_001033457.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002073.mm.1 NM_001033457.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002074.mm.1 TC0500000272.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500002074.mm.1 NM_001033457.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500002074.mm.1 NM_001033457.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232834 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2795677 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2795677 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline