| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500022659.mm.1 |
NM_008066.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500022659.mm.1 |
NM_008066.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500022659.mm.1 |
TC0500002559.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500022660.mm.1 |
NM_008066.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500022660.mm.1 |
NM_008066.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500022660.mm.1 |
TC0500002559.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500022661.mm.1 |
NM_008066.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500022661.mm.1 |
NM_008066.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500022661.mm.1 |
TC0500002559.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500022658.mm.1 |
NM_008066.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500022658.mm.1 |
NM_008066.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500022658.mm.1 |
TC0500002559.mm.1 |
MTA TC |