Search
Search

Limch1| Gene Symbol: Limch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001980.2 | NP_001001980.2 | 9 - 10 | 1043 | 73 | |
| NM_001256122.1 | NP_001243051.1 | 4 - 5 | 805 | 73 | ||
| GenBank mRNA | AK019457.1 | - | 4 - 5 | 662 | 73 | |
| AK122443.1 | - | 2 - 3 | 158 | 73 | ||
| AK134095.1 | - | 4 - 5 | 606 | 73 | ||
| AK147547.1 | - | 4 - 5 | 806 | 73 | ||
| CD804084.1 | - | 4 - 5 | 503 | 73 | ||
Set Membership: |
|