Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01211875_m1
          See other INSR GE Assays ›
          Gene Symbol:
          Insr
          Gene Name:
          insulin receptor
          Gene Aliases:
          4932439J01Rik, CD220, D630014A15Rik, IR, IR-A, IR-B
          Chromosome Location:
          Chr.8: 3150922 - 3279649 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01211875_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Insr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010568.2 NP_034698.2 1 - 2 588 57
          GenBank mRNA AK052187.1 - 1 - 2 523 57
          BY733217.1 - 1 - 2 590 57
          J05149.1 - 1 - 2 147 57
          KC356871.1 - 1 - 2 99 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001608.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Insr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          heart morphogenesis
          regulation of transcription, DNA-templated
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cell proliferation
          insulin receptor signaling pathway
          epidermis development
          organ morphogenesis
          regulation of hydrogen peroxide metabolic process
          positive regulation of glycoprotein biosynthetic process
          negative regulation of gene expression
          phosphorylation
          peptidyl-tyrosine phosphorylation
          transformation of host cell by virus
          signal transduction by protein phosphorylation
          male sex determination
          positive regulation of cell migration
          exocrine pancreas development
          activation of protein kinase activity
          activation of protein kinase B activity
          negative regulation of transporter activity
          cellular response to insulin stimulus
          response to tumor necrosis factor
          intracellular signal transduction
          peptidyl-tyrosine autophosphorylation
          positive regulation of phosphorylation
          glucose homeostasis
          positive regulation of MAPK cascade
          positive regulation of nitric oxide biosynthetic process
          positive regulation of glycogen biosynthetic process
          positive regulation of DNA replication
          positive regulation of glycolytic process
          positive regulation of mitotic nuclear division
          regulation of embryonic development
          positive regulation of glucose import
          protein autophosphorylation
          positive regulation of developmental growth
          protein heterotetramerization
          positive regulation of protein kinase B signaling
          positive regulation of respiratory burst
          cellular response to growth factor stimulus
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          insulin-activated receptor activity
          insulin-like growth factor receptor binding
          protein binding
          ATP binding
          GTP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          protein phosphatase binding
          protein domain specific binding
          lipoic acid binding
          insulin-like growth factor I binding
          insulin-like growth factor II binding
          protein complex binding
          3-phosphoinositide-dependent protein kinase binding
          phosphatidylinositol 3-kinase binding
          insulin binding
          insulin receptor substrate binding
          PTB domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800013194.mm.1 NM_010568.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013194.mm.1 TC0800001608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013195.mm.1 NM_010568.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013195.mm.1 TC0800001608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013193.mm.1 NM_010568.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013193.mm.1 TC0800001608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013191.mm.1 NM_010568.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013191.mm.1 TC0800001608.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline