Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kat6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081149.1 | NP_001074618.1 | 9 - 10 | 1969 | 110 | |
GenBank mRNA | AK028058.1 | - | 8 - 9 | 1888 | 110 | |
AK036885.1 | - | 9 - 10 | 1964 | 110 | ||
AK082463.1 | - | 9 - 10 | 1972 | 110 | ||
AK132474.1 | - | 8 - 9 | 1877 | 110 | ||
AK143408.1 | - | 9 - 10 | 1960 | 110 | ||
BC024786.1 | - | 1 - 2 | 95 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |