Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212031_m1
          See other OCIAD2 GE Assays ›
          Gene Symbol:
          Ociad2
          Gene Name:
          OCIA domain containing 2
          Gene Aliases:
          1810027I20Rik
          Chromosome Location:
          Chr.5: 73322199 - 73339355 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212031_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ociad2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026950.3 NP_081226.1 6 - 7 549 68
          GenBank mRNA AK007617.1 - 6 - 7 503 68
          BC044883.1 - 6 - 7 468 4971755 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002592.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ociad2

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128511 NM_026950.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765740 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888743 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61600 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139939 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158674 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128511 NM_026950.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23797131 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158674 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139939 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01132888 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01132901 - -
          Whole Mouse Genome G4122A A_51_P338886 BC044883.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338886 NM_026950.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338886 AK007617.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136995 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158674 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04129582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04129583 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04129584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23796973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23796972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23796977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23796979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23796974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23796983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23796978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23797077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04874461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04442335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04442342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04498622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04129583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04930054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04442344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04264725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04930052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04498624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04240261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04129582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04974173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04498631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04853952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04442341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04442345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04711261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04368895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04442331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04498630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23797077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04991712 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10530619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530620 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530624 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530622 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530623 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530617 - -
          Affymetrix HT MG-430 PM Array Plate 1435917_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453187_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10530615 - -
          Mouse Genome 430 2.0 Array 1435917_PM_at - -
          Mouse Genome 430 2.0 Array 1435917_at - -
          Mouse Genome 430 2.0 Array 1453187_PM_at - -
          Mouse Genome 430 2.0 Array 1453187_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433904_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433904_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500022994.mm.1 TC0500002592.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022994.mm.1 NM_026950.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022995.mm.1 TC0500002592.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022995.mm.1 NM_026950.4 Refseq
          Mouse Expression Set 430 1453187_at - -
          Mouse Expression Set 430 1435917_at - -
          Mouse Expression Set 430 1435917_PM_at - -
          Mouse Expression Set 430 1453187_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453187_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435917_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453187_at - -
          Affymetrix HT Mouse Genome 430B Array 1435917_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 363517 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105270403 - -
          Illumina mouse-6 v1.0 expression beadchip 105270403 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2482178 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2766875 - -
          mouse-6 v1.1 (Illumina) scl0004073.1_36-S - -
          Illumina Sentrix mouse-6.v1 bead array 105270403 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105270403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2943722 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2766875 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2943722 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0