| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900001228.mm.1 |
NM_194348.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001228.mm.1 |
NM_194348.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001228.mm.1 |
TC1900000108.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001230.mm.1 |
NM_194348.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001230.mm.1 |
NM_194348.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001230.mm.1 |
TC1900000108.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001229.mm.1 |
NM_194348.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001229.mm.1 |
NM_194348.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001229.mm.1 |
TC1900000108.mm.1 |
MTA TC |