Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212322_g1
          See other LYRM2 GE Assays ›
          Gene Symbol:
          Lyrm2
          Gene Name:
          LYR motif containing 2
          Gene Aliases:
          2610208E05Rik
          Chromosome Location:
          Chr.4: 32800259 - 32802268 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212322_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyrm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175364.4 NP_780573.1 2 - 3 216 84
          GenBank mRNA AK044791.1 - 2 - 3 211 84
          BC028533.1 - 2 - 3 198 1446940 84
          BY131987.1 - 2 - 3 216 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lyrm2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823558 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823554 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823552 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823553 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P75146 AK044791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P75146 BC028533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P75146 NM_175364.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P75146 AK044791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P75146 BC028533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P75146 NM_175364.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916031 AK018316.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916031 AK030007.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916031 AK083291.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916031 AK088917.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916031 AY428571.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916031 AY585206.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916031 BC026482.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916031 BC055320.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916031 NM_175238.5 refseq_mrna_mouse_NM
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137328 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P75146 AK044791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P75146 BC028533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P75146 NM_175364.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P75146 AK044791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P75146 BC028533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P75146 NM_175364.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22823540 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137328 - -
          Whole Mouse Genome G4122A A_52_P75146 AK044791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P75146 BC028533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P75146 NM_175364.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P75146 AK044791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P75146 BC028533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P75146 NM_175364.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122374 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137328 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03387993 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03577223 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03577218 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03387984 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03577220 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03577207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823552 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823558 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22823554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22823563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03577211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03577207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03577223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22823552 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10503829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10503830 - -
          Affymetrix HT MG-430 PM Array Plate 1427935_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427934_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10503828 - -
          Mouse Genome 430 2.0 Array 1427934_at - -
          Mouse Genome 430 2.0 Array 1427935_at - -
          Mouse Genome 430 2.0 Array 1427934_PM_at - -
          Mouse Genome 430 2.0 Array 1427935_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108755_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108755_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400001767.mm.1 NM_175364.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001767.mm.1 NM_175364.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001767.mm.1 TC0400000272.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400001770.mm.1 NM_175364.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001770.mm.1 NM_175364.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001770.mm.1 TC0400000272.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400001768.mm.1 NM_175364.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001768.mm.1 NM_175364.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400001768.mm.1 TC0400000272.mm.1 MTA TC
          Mouse Expression Set 430 1427935_at - -
          Mouse Expression Set 430 1427934_PM_at - -
          Mouse Expression Set 430 1427934_at - -
          Mouse Expression Set 430 1427935_PM_at - -
          Murine Genome U74v2 Set 102368_at - -
          Murine Genome U74v2 Set 102368_PM_at - -
          Mouse Genome 430A 2.0 Array 1427934_at - -
          Mouse Genome 430A 2.0 Array 1427935_PM_at - -
          Mouse Genome 430A 2.0 Array 1427935_at - -
          Mouse Genome 430A 2.0 Array 1427934_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427934_at - -
          Affymetrix HT Mouse Genome 430A Array 1427934_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427935_at - -
          Affymetrix HT Mouse Genome 430A Array 1427935_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 372374 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2450204 - -
          MouseWG-6 v2 Expression BeadChip 380021 - -
          Illumina mouse-6 v1.0 expression beadchip 2450204 - -
          Illumina mouse-6 v1.0 expression beadchip 380021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000380021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2739340 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2732585 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2739340 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2732585 - -
          mouse-6 v1.1 (Illumina) scl00108755.1_37-S - -
          mouse-6 v1.1 (Illumina) scl0108755.3_138-S - -
          Illumina Sentrix mouse-6.v1 bead array 380021 - -
          Illumina Sentrix mouse-6.v1 bead array 2450204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00108755.1_37-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0108755.3_138-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450204 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 380021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2806880 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2739340 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2732585 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2732585 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2806880 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2739340 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00108755.1_37-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline