| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800013310.mm.1 |
NM_029465.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013310.mm.1 |
NM_029465.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013310.mm.1 |
TC0800001617.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800013311.mm.1 |
NM_029465.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013311.mm.1 |
NM_029465.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013311.mm.1 |
TC0800001617.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800013314.mm.1 |
NM_029465.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013314.mm.1 |
NM_029465.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013314.mm.1 |
TC0800001617.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800013308.mm.1 |
NM_029465.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013308.mm.1 |
NM_029465.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013308.mm.1 |
TC0800001617.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800013313.mm.1 |
NM_029465.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013313.mm.1 |
NM_029465.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013313.mm.1 |
TC0800001617.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800013315.mm.1 |
NM_029465.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013315.mm.1 |
NM_029465.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013315.mm.1 |
TC0800001617.mm.1 |
MTA TC |