Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212488_m1
          See other MCEMP1 GE Assays ›
          Gene Symbol:
          Mcemp1
          Gene Name:
          mast cell expressed membrane protein 1
          Gene Aliases:
          1810033B17Rik
          Chromosome Location:
          Chr.8: 3665762 - 3668917 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212488_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mcemp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026985.1 NP_081261.1 5 - 6 506 66
          GenBank mRNA AK007679.1 - 5 - 6 514 66
          AK036449.1 - 5 - 6 507 66
          AK087121.1 - 5 - 6 512 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000020.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325856 AK087121.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325856 AK036449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325856 NM_026985.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325856 AK087121.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325856 AK036449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325856 NM_026985.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55962 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128954 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325856 AK087121.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325856 AK036449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325856 NM_026985.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325856 AK087121.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325856 AK036449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325856 NM_026985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497235 AK087121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497235 AK036449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497235 NM_026985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497235 AK007679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325856 AK087121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325856 AK036449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325856 NM_026985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325856 AK087121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325856 AK036449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325856 NM_026985.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07019786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07019785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07369036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07002329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07284906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615082 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569881 - -
          Affymetrix HT MG-430 PM Array Plate 1430612_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569877 - -
          Mouse Genome 430 2.0 Array 1430612_PM_at - -
          Mouse Genome 430 2.0 Array 1430612_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69189_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69189_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800000281.mm.1 NM_026985.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000281.mm.1 NM_026985.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000281.mm.1 TC0800000020.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000280.mm.1 NM_026985.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000280.mm.1 NM_026985.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000280.mm.1 TC0800000020.mm.1 MTA TC
          Mouse Expression Set 430 1430612_PM_at - -
          Mouse Expression Set 430 1430612_at - -
          Affymetrix HT Mouse Genome 430B Array 1430612_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430612_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 541638 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4010037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2712151 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2712151 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0