Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01213632_s1
          See other CLDN34B4 GE Assays ›
          Gene Symbol:
          Cldn34b4
          Gene Name:
          claudin 34B4
          Gene Aliases:
          4930428D18Rik
          Chromosome Location:
          Chr.X: 76385323 - 76397979 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01213632_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cldn34b4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033799.2 NP_001028971.1 3 814 119
          GenBank mRNA AK132827.1 - 3 815 119
          BC147626.1 - 3 684 9056616 119
          BC147629.1 - 3 712 9056619 119
          BC172110.1 - 3 684 9054396 119

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140973 NM_001033799.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140973 AK132827.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64447 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108925 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140973 NM_001033799.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140973 AK132827.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108925 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32475191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15102350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14887790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15106239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14709500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15176229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14691282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32475158 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10600542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600544 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600543 - -
          Affymetrix Mouse Gene 1.1 ST Array 10600541 - -
          Affymetrix Mouse Clariom S Array PSR0X00005216.mm.1 NM_001033799.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005216.mm.1 NM_001033799.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005216.mm.1 TC0X00000760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005209.mm.1 NM_001033799.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005209.mm.1 NM_001033799.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005209.mm.1 TC0X00000760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005213.mm.1 NM_001033799.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005213.mm.1 NM_001033799.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005213.mm.1 TC0X00000760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005211.mm.1 NM_001033799.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005211.mm.1 NM_001033799.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005211.mm.1 TC0X00000760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005215.mm.1 NM_001033799.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005215.mm.1 NM_001033799.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005215.mm.1 TC0X00000760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005210.mm.1 NM_001033799.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005210.mm.1 NM_001033799.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005210.mm.1 TC0X00000760.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 666041 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160886 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160886 - -
          mouse-6 v1.1 (Illumina) GI_85702128-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_2811775 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2811775 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0