Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01213635_m1
          See other GM14762 GE Assays ›
          Gene Symbol:
          Gm14762
          Gene Name:
          predicted gene 14762
          Gene Aliases:
          OTTMUSG00000017918
          Chromosome Location:
          Chr.X: 85825236 - 85827656 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01213635_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm14762
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK035200.1 - 214 60

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148674 AK052035.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174466 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148679 AK035200.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148674 AK052035.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15152919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32507363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14661187 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102680288 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220399 - -
          Illumina Sentrix mouse-6.v1 bead array 102680288 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102680288 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220399 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline