Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01214946_s1
          See other CPXCR1 GE Assays ›
          Gene Symbol:
          Cpxcr1
          Gene Name:
          CPX chromosome region, candidate 1
          Gene Aliases:
          Gm1143
          Chromosome Location:
          Chr.X: 116448944 - 116478783 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01214946_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cpxcr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033471.3 NP_001028643.2 3 1441 138
          GenBank mRNA AK133001.1 - 3 1443 138
          AV270501.2 - 1 460 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125439 AK133001.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125439 BC147520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125439 BC147548.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125439 NM_001033471.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275629 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172678 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P125439 AK133001.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P125439 BC147520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P125439 BC147548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P125439 NM_001033471.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03743002 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09400257 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09400251 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03743002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32616117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14794619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14794622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14794615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14788341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14788343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15137488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32616071 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10601542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601543 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601544 - -
          Affymetrix Mouse Gene 1.1 ST Array 10601541 - -
          Affymetrix Mouse Clariom S Array PSR0X00007483.mm.1 NM_001033471.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007483.mm.1 NM_001033471.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007483.mm.1 TC0X00001158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007486.mm.1 NM_001033471.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007486.mm.1 NM_001033471.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007486.mm.1 TC0X00001158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007484.mm.1 NM_001033471.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007484.mm.1 NM_001033471.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007484.mm.1 TC0X00001158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007487.mm.1 NM_001033471.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007487.mm.1 NM_001033471.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007487.mm.1 TC0X00001158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00007485.mm.1 NM_001033471.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007485.mm.1 NM_001033471.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00007485.mm.1 TC0X00001158.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 650219 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2846457 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2846457 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0